MilliporeSigma
  • Co-administration of human adipose-derived stem cells and low-level laser to alleviate neuropathic pain after experimental spinal cord injury.
Co-administration of human adipose-derived stem cells and low-level laser to alleviate neuropathic pain after experimental spinal cord injury.
Stem cell research & therapy (2019-06-27)
Arash Sarveazad, Atousa Janzadeh, Gholamreza Taheripak, Sima Dameni, Mahmoud Yousefifard, Farinaz Nasirinezhad
ABSTRACT
Evidence has suggested that human adipose-derived stem cells (hADSCs) and low-level laser has neuroprotective effects on spinal cord injury (SCI). Therefore, the combined effect of the hADSCs and laser on neuregeneration and neuropathic pain after SCI was investigated. Forty-eight adult male Wistar rats with 200-250 g weight were used. Thirty minutes after compression, injury with laser was irritated, and 1 week following SCI, about 1 × 106 cells were transplanted into the spinal cord. Motor function and neuropathic pain were assessed weekly. Molecular and histological studies were done at the end of the fourth week. The combined application of hADSCs and laser has significantly improved motor function recovery (p = 0.0001), hyperalgesia (p < 0.05), and allodynia (p < 0.05). GDNF mRNA expression was significantly increased in hADSCs and laser+hADSC-treated animals (p < 0.001). Finally, co-administration of hADSCs and laser has enhanced the number of axons around cavity more than other treatments (p < 0.001). The results showed that the combination of laser and ADSCs could significantly improve the motor function and alleviate SCI-induced allodynia and hyperalgesia. Therefore, using a combination of laser and hADSCs in future experimental and translational clinical studies is suggested.