MilliporeSigma
  • Characterization of an intestine-specific GH receptor knockout (IntGHRKO) mouse.
Characterization of an intestine-specific GH receptor knockout (IntGHRKO) mouse.
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society (2019-05-13)
Jonathan A Young, Elizabeth A Jensen, Austin Stevens, Silvana Duran-Ortiz, Edward O List, Darlene E Berryman, John J Kopchick
ABSTRACT
Growth hormone (GH) has been reported to enhance the intestinal barrier; as such, recombinant GH has been administered for several intestinal diseases. However, excess GH action has been implicated in increasing the risk of intestinal dysfunction. The goal of this study was to examine the direct effects of GH on the small and large intestines to clarify the role GH plays in intestinal function through the use of a mouse model. An intestinal epithelial-specific GH receptor (GHR) knockout (IntGHRKO) mouse line was generated using Cre-lox with the villin promoter driving Cre expression. The generated mice were characterized with respect to growth and intestinal phenotypes. IntGHRKO mice showed no significant changes in body length, weight, or composition compared to floxed controls. Male IntGHRKO mice had significantly shorter large intestines at 4 and 12 months of age. Intestinal barrier function was assessed by measuring the expression of tight junction related genes, as well as levels of serum endotoxin and fecal albumin. Results showed sex differences as males had an increase in occludin levels but normal serum endotoxin and fecal albumin; while, females had changes in fecal albumin levels with normal occludin and serum endotoxin. Evaluation of glucose tolerance and fat absorption also showed sex differences as females were glucose intolerant, while males had impaired fat absorption. Histopathology revealed a trend towards decreased villus height in males, which could explain the sex difference in glucose homeostasis. Overall, the data demonstrate that disruption of GH on the intestinal epithelial cells modestly affects the intestinal gross anatomy, morphology, and function in a sex-specific manner.