MilliporeSigma
  • Isochorismate-derived biosynthesis of the plant stress hormone salicylic acid.
Isochorismate-derived biosynthesis of the plant stress hormone salicylic acid.
Science (New York, N.Y.) (2019-08-03)
Dmitrij Rekhter, Daniel Lüdke, Yuli Ding, Kirstin Feussner, Krzysztof Zienkiewicz, Volker Lipka, Marcel Wiermer, Yuelin Zhang, Ivo Feussner
ABSTRACT
The phytohormone salicylic acid (SA) controls biotic and abiotic plant stress responses. Plastid-produced chorismate is a branch-point metabolite for SA biosynthesis. Most pathogen-induced SA derives from isochorismate, which is generated from chorismate by the catalytic activity of ISOCHORISMATE SYNTHASE1. Here, we ask how and in which cellular compartment isochorismate is converted to SA. We show that in Arabidopsis, the pathway downstream of isochorismate requires only two additional proteins: ENHANCED DISEASE SUSCEPTIBILITY5, which exports isochorismate from the plastid to the cytosol, and the cytosolic amidotransferase avrPphB SUSCEPTIBLE3 (PBS3). PBS3 catalyzes the conjugation of glutamate to isochorismate to produce isochorismate-9-glutamate, which spontaneously decomposes into SA and 2-hydroxy-acryloyl-N-glutamate. The minimal requirement of three compartmentalized proteins controlling unidirectional forward flux may protect the pathway against evolutionary forces and pathogen perturbations.