MilliporeSigma
Gas-Phase Synthesis of Triphenylene (C18 H12 ).
Chemphyschem : a European journal of chemical physics and physical chemistry (2019-02-03)
Long Zhao, Bo Xu, Utuq Ablikim, Wenchao Lu, Musahid Ahmed, Mikhail M Evseev, Eugene K Bashkirov, Valeriy N Azyazov, A Hasan Howlader, Stanislaw F Wnuk, Alexander M Mebel, Ralf I Kaiser
ABSTRACT
For the last decades, the hydrogen-abstraction-acetylene-addition (HACA) mechanism has been widely invoked to rationalize the high-temperature synthesis of PAHs as detected in carbonaceous meteorites (CM) and proposed to exist in the interstellar medium (ISM). By unravelling the chemistry of the 9-phenanthrenyl radical ([C14 H9 ]. ) with vinylacetylene (C4 H4 ), we present the first compelling evidence of a barrier-less pathway leading to a prototype tetracyclic PAH - triphenylene (C18 H12 ) - via an unconventional hydrogen abstraction-vinylacetylene addition (HAVA) mechanism operational at temperatures as low as 10 K. The barrier-less, exoergic nature of the reaction reveals HAVA as a versatile reaction mechanism that may drive molecular mass growth processes to PAHs and even two-dimensional, graphene-type nanostructures in cold environments in deep space thus leading to a better understanding of the carbon chemistry in our universe through the untangling of elementary reactions on the most fundamental level.