MilliporeSigma
  • CD1b presents self and Borrelia burgdorferi diacylglycerols to human T cells.
CD1b presents self and Borrelia burgdorferi diacylglycerols to human T cells.
European journal of immunology (2019-03-12)
Peter Reinink, Michael N T Souter, Tan-Yun Cheng, Tamara van Gorkom, Stefanie Lenz, Joanna Kubler-Kielb, Klemen Strle, Kristin Kremer, Steven F T Thijsen, Allen C Steere, Dale I Godfrey, Daniel G Pellicci, D Branch Moody, Ildiko Van Rhijn
ABSTRACT
Lyme disease is a common multisystem disease caused by infection with a tick-transmitted spirochete, Borrelia burgdorferi and related Borrelia species. The monoglycosylated diacylglycerol known as B. burgdorferi glycolipid II (BbGL-II) is a major target of antibodies in sera from infected individuals. Here, we show that CD1b presents BbGL-II to human T cells and that the TCR mediates the recognition. However, we did not detect increased frequency of CD1b-BbGL-II binding T cells in the peripheral blood of Lyme disease patients compared to controls. Unexpectedly, mapping the T cell specificity for BbGL-II-like molecules using tetramers and activation assays revealed a concomitant response to CD1b-expressing APCs in absence of BbGL-II. Further, among all major classes of self-lipid tested, BbGL-II responsive TCRs show strong cross-reactivity to diacylglycerol, a self-lipid antigen with structural similarities to BbGL-II. Extending prior work on MHC and CD1b, CD1c, and CD1d proteins, this study provides evidence for cross-reactive CD1b-restricted T cell responses to bacterial and self-antigens, and identifies chemically defined targets for future discovery of self and foreign antigen cross-reactive T cells.