MilliporeSigma
  • Synthesis of Novel Benzimidazole-2-carboxamide Derivatives and in Vivo Antihyperlipidemic Activity Evaluation.
Synthesis of Novel Benzimidazole-2-carboxamide Derivatives and in Vivo Antihyperlipidemic Activity Evaluation.
Chemical & pharmaceutical bulletin (2018-04-03)
Ghassan Abu Sheikha, Majdi Mohammad Bkhaitan, Hanin Kalloush, Lama Hamadneh, Reema Abu Khalaf, Tariq Al-Qirim, Yusuf Al-Hiari
ABSTRACT
Hyperlipidemia is known as an elevation of plasma lipid components. It contributes significantly to atherosclerosis which is one of the most important causative factors in cardiovascular diseases. Agents that cause a dramatic decrease in serum lipid levels are of great value in the treatment of cardiovascular diseases. For this purpose, a new series of benzimidazole propyl carboxamide benzophenone derivatives have been synthesized (7, 8, and 9). These compounds were tested in vivo to evaluate their potential hypolipidemic activity using Triton WR-1339 induced hyperlipidemic rats. All the synthesized compounds have proved to be highly biologically active, with compound 9 being the most active derivative.