MilliporeSigma
  • 1,8-Cineole blocks voltage-gated L-type calcium channels in tracheal smooth muscle.
1,8-Cineole blocks voltage-gated L-type calcium channels in tracheal smooth muscle.
Pflugers Archiv : European journal of physiology (2018-09-13)
Átila Pereira-Gonçalves, Francisco Walber Ferreira-da-Silva, Camille Maria de Holanda-Angelin-Alves, Ana Carolina Cardoso-Teixeira, Andrelina Noronha Coelho-de-Souza, José Henrique Leal-Cardoso
ABSTRACT
1,8-Cineole is a cyclic monoterpenoid used in folk medicine for treatment of numerous respiratory diseases and other infections. 1,8-Cineole has anti-inflammatory, antioxidant, and myorelaxant effects, as well as low toxicity. In the present study, the effects of 1,8-cineole on contractility and voltage-gated calcium channels (VGCC) in tracheal smooth muscle were investigated. Intact and dissociated tracheal smooth muscle were used for muscle contraction and patch-clamp recordings, respectively. In experiments involving muscle contraction, 1,8-cineole potentiated contractions at low concentrations and relaxed contractions induced by isotonic K+ at high concentrations. AMTB (a TRPM8 channel blocker) reduced the potentiation induced by 1,8-cineole while indomethacin (a COX inhibitor) did not block this effect. In dissociated myocytes, 1,8-cineole partially blocked Ba2+ currents through VGCC in a concentration-dependent manner. 1,8-Cineole shifted the steady-state activation and inactivation curves to the left and also reduced the current decay time constant. In conclusion, 1,8-cineole has a dual effect on tracheal smooth muscle contraction resulting in a biphasic effect. Our data suggest that the potentiation effect is mediated by activation of TRPM8 channels and the relaxation effect is mediated by the blockage of L-type VGCC.