MilliporeSigma
  • Stakeholder Perceptions of the Barriers to Receiving and Delivering Exposure-Based Cognitive Behavioral Therapy for Anxiety Disorders in Adult Community Mental Health Settings.
Stakeholder Perceptions of the Barriers to Receiving and Delivering Exposure-Based Cognitive Behavioral Therapy for Anxiety Disorders in Adult Community Mental Health Settings.
Community mental health journal (2018-03-07)
Kate Wolitzky-Taylor, Bowen Chung, Sarah Kate Bearman, Joanna Arch, Jason Grossman, Karissa Fenwick, Rebecca Lengnick-Hall, Jeanne Miranda
ABSTRACT
CBT is considered the first-line treatment for anxiety disorders, particularly when it involves gradual confrontation with feared stimuli (i.e., exposure); however, delivery of CBT for anxiety disorders in real-world community clinics is lacking. This study utilized surveys we developed with key stakeholder feedback (patient, provider, and administrator) to assess patient and provider/administrator perceptions of the barriers to delivering (or receiving) CBT for anxiety disorders. Providers/administrators from two counties in California (N = 106) indicated lack of training/competency as primary barriers. Patients in one large county (N = 42) reported their own symptoms most often impacted treatment receipt. Both groups endorsed acceptability of exposure but indicated that its use in treatment provided/received had been limited. Implications and recommendations are discussed.