MilliporeSigma
  • Palladium Catalyst with Task-Specific Ionic Liquid Ligands: Intracellular Reactions and Mitochondrial Imaging with Benzothiadiazole Derivatives.
Palladium Catalyst with Task-Specific Ionic Liquid Ligands: Intracellular Reactions and Mitochondrial Imaging with Benzothiadiazole Derivatives.
The Journal of organic chemistry (2019-04-09)
Thiago O Carvalho, Pedro H P R Carvalho, Jose R Correa, Bruna C Guido, Gisele A Medeiros, Marcos N Eberlin, Sara E Coelho, Josiel B Domingos, Brenno A D Neto
ABSTRACT
A water-soluble and charge-tagged palladium complex (PdMAI) was found to function inside breast cancer live cells of the MCF-7 lineage as an efficient catalyst for cross-coupling reaction. PdMAI, bearing two ionophilic task-specific ionic liquids as ligands, efficiently catalyzed both in cellulo Suzuki and Buchwald-Hartwig amination reactions. For the first time, therefore, the Buchwald-Hartwig amination is described to occur inside the highly complex cellular environment. The 2,1,3-benzothiadiazole (BTD) core was used as the base for the syntheses, and two π-extended fluorescent derivatives (BTD-2APy) and (BTD-1AN), which were found to emit in the green and red channels, had impressive mitochondrial affinity. These chromophores allowed for selective mitochondrial imaging and tracking.