MilliporeSigma
  • Farnesoid X receptor agonist GW4064 indirectly inhibits HCV entry into cells via down-regulating scavenger receptor class B type I.
Farnesoid X receptor agonist GW4064 indirectly inhibits HCV entry into cells via down-regulating scavenger receptor class B type I.
European journal of pharmacology (2019-03-25)
Zhou-Yi Wu, Hu Li, Jian-Rui Li, Xiao-Qin Lv, Jian-Dong Jiang, Zong-Gen Peng
ABSTRACT
Farnesoid X receptor (FXR) agonists play important regulatory roles in bile acid, lipid and glucose metabolism in vitro and in vivo. Thus, FXR agonists exhibit potential therapeutic effects on metabolism-related diseases that are associated with extrahepatic manifestations induced by hepatitis C virus (HCV) infection. This study investigated the effect and mechanism of FXR agonist GW4064 against HCV in vitro to explore the potential application of FXR agonists. Results showed that GW4064 and other FXR agonists have potent antiviral activity against HCV in Huh7.5 cells. GW4064 down-regulated the expression of scavenger receptor class B type I protein via FXR and thereby indirectly inhibited HCV entry into cells, leading to interruption of HCV life cycle. GW4064 also exhibited synergistic anti-HCV effect with known direct-acting antiviral agents (DAAs) used in the clinic and remained sensitive to DAA-resistant HCV mutations. Therefore, FXR agonists are also a kind of antiviral agent, and might be helpful in treatment of HCV-induced hepatic and extrahepatic manifestations.