MilliporeSigma
  • Injecting RNA interference lentiviruses targeting the muscarinic 3 receptor gene into the bladder wall inhibits neurogenic detrusor overactivity in rats with spinal cord injury.
Injecting RNA interference lentiviruses targeting the muscarinic 3 receptor gene into the bladder wall inhibits neurogenic detrusor overactivity in rats with spinal cord injury.
Neurourology and urodynamics (2018-12-15)
Zhenhua Shang, Chunsong Jia, Hao Yan, Bo Cui, Jiangtao Wu, Qi Wang, Wei Gao, Xin Cui, Jin Li, Tongwen Ou
ABSTRACT
To investigate the effects of injecting RNA interference (RNAi) lentiviruses targeting the muscarinic 3 (M3 ) receptor gene into the bladder wall on bladder activity in rats with spinal cord injury (SCI). Four M3 RNAi lentiviruses were constructed and used to infect primary cultured bladder smooth muscle cells (BSMCs). Western blotting and quantitative reverse transcription polymerase chain reaction (qRT-PCR) were performed to determine the optimal RNAi lentivirus with the highest interference efficiency. Female Wistar rats were subjected spinal cord transection at T9-10 and randomly divided into three groups (n = 8), namely, blank control, negative control, and experimental groups, and injected into the bladder wall with saline, negative control shRNA, and M3 RNAi lentiviruses, respectively, 1 week after spinal cord transection. The normal rats were used as normal control group. Urodynamic parameters and bladder tissues were evaluated in the different groups. An M3 RNAi lentivirus with the highest interference efficiency (78.9%) was constructed and identified. Three weeks after injecting M3 RNAi lentiviruses into the bladder wall, Western blotting and qRT-PCR showed that the M3 receptor was significantly downregulated in the experimental group. Cystometric evaluation suggested that downregulating M3 receptor expression could substantially decrease basal pressure, residual volume, and non-voiding contraction number, increase intercontraction interval, and significantly improve bladder compliance in rats with SCI. Injecting RNAi lentiviruses targeting the M3 receptor gene into the bladder wall could effectively inhibit neurogenic detrusor overactivity (NDO) due to SCI. Thus, this approach may be a potential treatment for NDO in SCI.