MilliporeSigma
A Neural Circuit for Gut-Induced Reward.
Cell (2018-09-25)
Wenfei Han, Luis A Tellez, Matthew H Perkins, Isaac O Perez, Taoran Qu, Jozelia Ferreira, Tatiana L Ferreira, Daniele Quinn, Zhong-Wu Liu, Xiao-Bing Gao, Melanie M Kaelberer, Diego V Bohórquez, Sara J Shammah-Lagnado, Guillaume de Lartigue, Ivan E de Araujo
ABSTRACT
The gut is now recognized as a major regulator of motivational and emotional states. However, the relevant gut-brain neuronal circuitry remains unknown. We show that optical activation of gut-innervating vagal sensory neurons recapitulates the hallmark effects of stimulating brain reward neurons. Specifically, right, but not left, vagal sensory ganglion activation sustained self-stimulation behavior, conditioned both flavor and place preferences, and induced dopamine release from Substantia nigra. Cell-specific transneuronal tracing revealed asymmetric ascending pathways of vagal origin throughout the CNS. In particular, transneuronal labeling identified the glutamatergic neurons of the dorsolateral parabrachial region as the obligatory relay linking the right vagal sensory ganglion to dopamine cells in Substantia nigra. Consistently, optical activation of parabrachio-nigral projections replicated the rewarding effects of right vagus excitation. Our findings establish the vagal gut-to-brain axis as an integral component of the neuronal reward pathway. They also suggest novel vagal stimulation approaches to affective disorders.
Sigma-Aldrich
Anti-Fluorescent Gold Antibody, from rabbit
Sigma-Aldrich
Metoclopramide hydrochloride, solid
Sigma-Aldrich
Clozapine