MilliporeSigma
  • Potentiation of serotonin signaling protects against intestinal ischemia and reperfusion injury in mice.
Potentiation of serotonin signaling protects against intestinal ischemia and reperfusion injury in mice.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2018-11-06)
John J Tackett, Neeru Gandotra, Michaela C Bamdad, Eleanor D Muise, Robert A Cowles
ABSTRACT
Knock-out of serotonin re-uptake transporters (SERT) or use of selective serotonin re-uptake inhibitors (SSRIs) potentiates enteric serotonin (5-HT) signaling and stimulates enterocyte proliferation. We hypothesized that increased serotonin signaling would mitigate epithelial injury from intestinal ischemia and reperfusion (I/R). Mice lacking SERT (SERTKO mice) and wild-type littermates (WTLM) were subjected to intestinal ischemia by superior mesenteric artery (SMA) occlusion. At intervals post-laparotomy with or without ischemia, ileum was harvested and prepared for staining. A WTLM subgroup treated with SSRI after SMA occlusion followed by reperfusion was also sacrificed and analyzed. Mucosal injury was scored, percentage of injured villi calculated, and enterocyte proliferation measured. Lastly, staining for enterocytes, enteroendocrine cells, and goblet cells, villus epithelial cellular make-up was investigated at baseline and 14 days after injury. Measurements were compared between groups using t test and chi-squared test. Mucosal injury after I/R was significantly decreased in SERTKO and SSRI-treated mice compared to WTLM at all intervals except baseline. Enterocyte proliferation was greater in SERTKO and SSRI-treated mice without alteration in cellular composition along villi (P > 0.05). Potentiation of 5-HT signaling is associated with mucosal protection from intestinal I/R injury without alterations in villus cell distribution, possibly via increased rates of enterocyte renewal.