MilliporeSigma
  • N-isobutyryl-His-Trp-Ala-Val-D-Ala-His-Leu-NHMe (ICI 216140) a potent in vivo antaconist analogue of bombesin/gastrin releasing peptide (BN/GRP) derived from the C-terminal sequence lacking the final methionine residue.
N-isobutyryl-His-Trp-Ala-Val-D-Ala-His-Leu-NHMe (ICI 216140) a potent in vivo antaconist analogue of bombesin/gastrin releasing peptide (BN/GRP) derived from the C-terminal sequence lacking the final methionine residue.
Life sciences (1989-01-01)
R Camble, R Cotton, A S Dutta, A Garner, C F Hayward, V E Moore, P B Scholes
ABSTRACT
The GRP receptor mediated growth response in Swiss 3T3 cells has been used to identify BN/GRP antagonists. Analysis of bombesin antagonism by substance P analogues and by truncated GRP analogues revealed that deletion of the C-terminal methionine residue was important for antagonism. Des-Met analogues showing potent antagonist activity in the in vitro 3T3 system (IC50 approximately 2nM) were synthesized. Further structural modification of these peptides led to the identification of (CH3)2CHCO-His-Trp-Ala-Val-D-Ala-His-Leu-NHCH3 (ICI 216140) which reduced bombesin-stimulated rat pancreatic amylase secretion to basal levels when administered subcutaneously at 2.0 mg per kg.