MilliporeSigma
  • Fibroblast-Mediated Immunoregulation of Macrophage Function Is Maintained after Irradiation.
Fibroblast-Mediated Immunoregulation of Macrophage Function Is Maintained after Irradiation.
Cancers (2019-05-22)
Rodrigo Berzaghi, Muhammad Asad Ahktar, Ashraful Islam, Brede D Pedersen, Turid Hellevik, Inigo Martinez-Zubiaurre
ABSTRACT
The abilities of cancer-associated fibroblasts (CAFs) to regulate immune responses in the context of radiotherapy remain largely unknown. This study was undertaken to determine whether ionizing radiation alters the CAF-mediated immunoregulatory effects on macrophages. CAFs were isolated from freshly-resected non-small cell lung cancer tumors, while monocyte-derived macrophages were prepared from peripheral blood of healthy donors. Experimental settings included both (CAF-macrophage) co-cultures and incubations of M0 and M1-macrophages in the presence of CAF-conditioned medium (CAF-CM). Functional assays to study macrophage polarization/activation included the expression of cell surface markers, production of nitric oxide, secretion of inflammatory cytokines and migratory capacity. We show that CAFs promote changes in M0-macrophages that harmonize with both M1-and M2-phenotypes. Additionally, CAFs inhibit pro-inflammatory features of M1-macrophages by reducing nitric oxide production, pro-inflammatory cytokines, migration, and M1-surface markers expression. Radiation delivered as single-high dose or in fractioned regimens did not modify the immunoregulatory features exerted by CAFs over macrophages in vitro. Protein expression analyses of CAF supernatants showed that irradiated and non-irradiated CAFs produce approximately the same protein levels of immunoregulators. Thus, CAF-derived soluble factors mediate measurable changes on uncommitted macrophages and down-regulate pro-inflammatory features of M1-polarized macrophages. Notably, ionizing radiation does not curtail the CAF-mediated immunosuppressive effects.