MilliporeSigma
  • NIR/pH dual-responsive polysaccharide-encapsulated gold nanorods for enhanced chemo-photothermal therapy of breast cancer.
NIR/pH dual-responsive polysaccharide-encapsulated gold nanorods for enhanced chemo-photothermal therapy of breast cancer.
Materials science & engineering. C, Materials for biological applications (2019-07-28)
Weijun Xu, Jinlei Wang, Junmin Qian, Guanghui Hou, Yaping Wang, Lijie Ji, Aili Suo
ABSTRACT
Combination of different therapy modalities with multiple tumoricidal mechanisms has emerged as a promising anticancer strategy. Herein, we reported an aldehyde/catechol-functionalized hyaluronic acid (DAHA) and hydroxyethyl chitosan (HECS) decorated gold nanorod (GNR) platform for combined chemo-photothermal therapy of breast cancer. The DAHA was synthesized by conjugating dopamine onto oxidized hyaluronic acid. The nanoplatform was prepared by successively modifying GNR with DAHA via Au-catechol bonds, conjugating DOX onto DAHA moiety through Schiff base linkage, and coating HECS interlayer for charge-reversal and hyaluronic acid corona for tumor cell targeting. The resulting nanoplatform GNR-HADOXCH exhibited acid-triggered surface charge-reversal and pH/NIR dual-responsive drug release behaviors. The nanoplatform could be efficiently internalized into MCF-7 breast cancer cells and displayed greater cancer cell killing than individual modalities. Therefore, polysaccharide decoration could ensure the co-delivery of GNR and DOX into cancer cells, and the developed GNR-HADOXCH holds great potential for breast cancer treatment.