MilliporeSigma
  • Unnatural spirocyclic oxindole alkaloids biosynthesis in Uncaria guianensis.
Unnatural spirocyclic oxindole alkaloids biosynthesis in Uncaria guianensis.
Scientific reports (2019-08-07)
Adriana A Lopes, Bianca Chioca, Bruno Musquiari, Eduardo J Crevelin, Suzelei de C França, Maria Fatima das G Fernandes da Silva, Ana Maria S Pereira
ABSTRACT
Spiro-oxindole scaffolds have been studied due to their promising therapeutic potential. In the Amazon rainforest there are two important Uncaria species known as "cat's claw", which biosynthesize spirocyclic oxindole alkaloids; Uncaria tomentosa (Willd. ex Schult.) DC. and Uncaria guianensis (Aublet) Gmell. We carried out a precursor-directed biosynthesis approach with U. guianensis and successfully obtained oxindole alkaloid analogues with molecular mass corresponding to the addition of a methyl or fluorine group on the oxindole ring using tryptamine analogue precursors. Two of these novel oxindole alkaloid analogues (3b-7-methyl-isomitraphylline and 3c-6-fluoro-isomitraphylline) were isolated and characterized by NMR spectroscopy and ESI-QTOF-MS/MS. Having established a substrate feeding protocol for these plantlets, the biosynthetic route for mitraphylline (1), rhynchophylline (2), isomitraphylline (3) and isorhynchophylline (4) was also investigated using 13C-precursors (1-13C-D-glucose, 2-13C-tryptophan, 1-13C-DL-glyceraldehyde, and methyl-13C-D-methionine).