MilliporeSigma
  • Liver-derived fibroblast growth factor 21 mediates effects of glucagon-like peptide-1 in attenuating hepatic glucose output.
Liver-derived fibroblast growth factor 21 mediates effects of glucagon-like peptide-1 in attenuating hepatic glucose output.
EBioMedicine (2019-03-05)
Junling Liu, Kun Yang, Jin Yang, Wenhua Xiao, Yunyi Le, Fei Yu, Liangbiao Gu, Shan Lang, Qing Tian, Tianru Jin, Rui Wei, Tianpei Hong
ABSTRACT
Glucagon-like peptide-1 (GLP-1) and its based agents improve glycemic control. Although their attenuating effect on hepatic glucose output has drawn our attention for decades, the potential mechanisms remain unclear. Cytokine array kit was used to assess cytokine profiles in db/db mice and mouse primary hepatocytes treated with exenatide (exendin-4). Two diabetic mouse models (db/db and Pax6m/+) were treated with a GLP-1 analog exenatide or liraglutide. The expression and secretion of fibroblast growth factor 21 (FGF21) in the livers of diabetic mice, primary mouse and human hepatocytes, and the human hepatic cell line HepG2 treated with or without GLP-1 analog were measured. Blockage of FGF21 with neutralizing antibody or siRNA, or hepatocytes isolated from Fgf21 knockout mice were used, and the expression and activity of key enzymes in gluconeogenesis were analyzed. Serum FGF21 level was evaluated in patients with type 2 diabetes (T2D) receiving exenatide treatment. Utilizing the cytokine array, we identified that FGF21 secretion was upregulated by exenatide (exendin-4). Similarly, FGF21 production in hepatocytes was stimulated by exenatide or liraglutide. FGF21 blockage attenuated the inhibitory effects of the GLP-1 analogs on hepatic glucose output. Similar results were also observed in primary hepatocytes from Fgf21 knockout mice. Furthermore, exenatide treatment increased serum FGF21 level in patients with T2D, particularly in those with better glucose control. We identify that function of GLP-1 in inhibiting hepatic glucose output is mediated via the liver hormone FGF21. Thus, we provide a new extra-pancreatic mechanism by which GLP-1 regulates glucose homeostasis. FUND: National Key Research and Development Program of China, the National Natural Science Foundation of China, the Natural Science Foundation of Beijing and Peking University Medicine Seed Fund for Interdisciplinary Research.