MilliporeSigma
  • Bioactivity of the Cymbopogon citratus (Poaceae) essential oil and its terpenoid constituents on the predatory bug, Podisus nigrispinus (Heteroptera: Pentatomidae).
Bioactivity of the Cymbopogon citratus (Poaceae) essential oil and its terpenoid constituents on the predatory bug, Podisus nigrispinus (Heteroptera: Pentatomidae).
Scientific reports (2019-06-09)
Bruno Pandelo Brügger, Luis Carlos Martínez, Angelica Plata-Rueda, Barbara Monteiro de Castro E Castro, Marcus Alvarenga Soares, Carlos Frederico Wilcken, Amélia Guimarães Carvalho, José Eduardo Serrão, José Cola Zanuncio
ABSTRACT
Podisus nigrispinus Dallas (Heteroptera: Pentatomidae), released in biological control programs, is a predator of Lepidopteran and Coleopteran species. Lemongrass essential oil and its constituents can be toxic to this natural enemy. The major constituents of lemongrass essential oil are neral (31.5%), citral (26.1%), and geranyl acetate (2.27%). Six concentrations of lemongrass essential oil and of its citral and geranyl acetate constituents were applied to the thorax of P. nigrispinus nymphs and adults. The walking and respiratory behavior of the P. nigrispinus third-instar nymphs, treated with citral and geranyl acetate at the LD50 and LD90 doses, were analyzed with video and respirometer. The lemongrass essential oil toxicity increased from first- to fifth-instar P. nigrispinus nymphs. The P. nigrispinus respiration rates (μL de CO2 h-1/insect) with citral and geranyl acetate in the LD50 and LD90 differed. Nymphs exposed to the lemongrass essential oil and its constituents on treated surfaces presented irritability or were repelled. Podisus nigrispinus adults were tolerant to the lemongrass essential oil and its constituents, geranyl acetate and citral. The altered respiratory activity with geranyl acetate and the fact that they were irritated and repelled by citral suggest caution with regard to the use of the lemongrass essential oil and its constituents in integrated pest management incorporating this predator, in order to avoid diminishing its efficiency against the pests.