MilliporeSigma
  • Common cellular origin and diverging developmental programs for different sesamoid bones.
Common cellular origin and diverging developmental programs for different sesamoid bones.
Development (Cambridge, England) (2019-02-13)
Shai Eyal, Sarah Rubin, Sharon Krief, Lihi Levin, Elazar Zelzer
ABSTRACT
Sesamoid bones are small auxiliary bones that form near joints and contribute to their stability and function. Thus far, providing a comprehensive developmental model or classification system for this highly diverse group of bones has been challenging. Here, we compare our previously reported mechanisms of patella development in the mouse with those of two anatomically different sesamoids, namely lateral fabella and digit sesamoids. We show that all three types of sesamoid bones originate from Sox9+ /Scx+ progenitors under the regulation of TGFβ and independently of mechanical stimuli from muscles. Whereas BMP2 regulates the growth of all examined sesamoids, the differentiation of lateral fabella or digit sesamoids is regulated redundantly by BMP4 and BMP2. Next, we show that whereas patella and digit sesamoids initially form in juxtaposition to long bones, lateral fabella forms independently and at a distance. Finally, our evidence suggests that, unlike the synovial joint that separates patella from femur, digit sesamoids detach from the phalanx by formation of a fibrocartilaginous joint. These findings highlight both common and divergent molecular and mechanical features of sesamoid bone development, which underscores their evolutionary plasticity.
Sigma-Aldrich
Proteinase K–Agarose from Tritirachium album, lyophilized powder
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Roche
Anti-Digoxigenin-POD, Fab fragments, from sheep