MilliporeSigma
  • A Novel Gold Nanoprobe for a Simple Electrochemiluminescence Determination of a Prostate-specific Antigen Based on a Peptide Cleavage Reaction.
A Novel Gold Nanoprobe for a Simple Electrochemiluminescence Determination of a Prostate-specific Antigen Based on a Peptide Cleavage Reaction.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2018-10-10)
Zhejian Li, Shumin Wang, Xuemei Fan, Baoyue Cao, Chunsheng Zhou
ABSTRACT
A novel gold nanoprobe for a sensitive and simple determination of a prostate-specific antigen (PSA) was designed on the basis of homogeneous detection and a peptide cleavage reaction. The gold nanoprobe (AuNPs-peptide-Ru1) consisted of a specific peptide tagged with a ruthenium(II) complex (Ru1) and gold nanoparticles (AuNPs) conjugated with the peptide via the strong Au-S bond between the AuNPs surface and the thiol group of the peptide. The electrochemiluminescence (ECL) enzymatic-cleavage-reaction-based bioanalytic system based on homogeneous detection has overcome shortcomings from a complicated fabrication process of traditional electrodes. In the presence of the target PSA, it specifically cleaved the peptide of the AuNPs-peptide-Ru1, and the ECL signal substance (Ru1) part dissociated from AuNPs-peptide-Ru1. This resulted in an increase in the ECL intensity. The ECL biosensor could detect PSA concentrations in the range from 1.0 × 10-12 to 1.0 × 10-9 g/mL, the detection limit was 4.0 × 10-13 g/mL. The assay with the advantages of a simple method for PSA was selective and fast. It is superior to the immunoassay, and is a promising strategy to develop biosensors based on enzymatic cleavage including electrochemistry and optics.