MilliporeSigma
  • Harnessing Androgen Receptor Pathway Activation for Targeted Alpha Particle Radioimmunotherapy of Breast Cancer.
Harnessing Androgen Receptor Pathway Activation for Targeted Alpha Particle Radioimmunotherapy of Breast Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2018-09-27)
Daniel L J Thorek, Anson T Ku, Nicholas Mitsiades, Darren Veach, Philip A Watson, Dipti Metha, Sven-Erik Strand, Sai Kiran Sharma, Jason S Lewis, Diane S Abou, Hans G Lilja, Steven M Larson, Michael R McDevitt, David Ulmert
ABSTRACT
The impact of androgen receptor (AR) activity in breast cancer biology is unclear. We characterized and tested a novel therapy to an AR-governed target in breast cancer.Experimental Design: We evaluated the expression of prototypical AR gene products human kallikrein 2 (hK2) and PSA in breast cancer models. We screened 13 well-characterized breast cancer cell lines for hK2 and PSA production upon in vitro hormone stimulation by testosterone [dihydrotestosterone (DHT)]. AR-positive lines were further evaluated by exposure to estrogen (17β-Estradiol) and the synthetic progestin D-Norgestrel. We then evaluated an anti-hK2-targeted radiotherapy platform (hu11B6), labeled with alpha (α)-particle emitting Actinium-225, to specifically treat AR-expressing breast cancer xenografts under hormone stimulation. D-Norgestrel and DHT activated the AR pathway, while 17β-Estradiol did not. Competitive binding for AR protein showed similar affinity between DHT and D-Norgestrel, indicating direct AR-ligand interaction. In vivo production of hK2 was sufficient to achieve site-specific delivery of therapeutic radionuclide to tumor tissue at >20-fold over background muscle uptake; effecting long-term local tumor control. [225Ac]hu11B6 targeted radiotherapy was potentiated by DHT and by D-Norgestrel in murine xenograft models of breast cancer. AR activity in breast cancer correlates with kallikrein-related peptidase-2 and can be activated by D-Norgestrel, a common contraceptive, and AR induction can be harnessed for hK2-targeted breast cancer α-emitter radiotherapy.