MilliporeSigma
  • Rev-erbα can regulate the NF-κB/NALP3 pathway to modulate lipopolysaccharide-induced acute lung injury and inflammation.
Rev-erbα can regulate the NF-κB/NALP3 pathway to modulate lipopolysaccharide-induced acute lung injury and inflammation.
International immunopharmacology (2019-05-28)
Dapeng Yu, Xiangzhi Fang, Yingying Xu, Huashi Xiao, Tianfeng Huang, Yang Zhang, Yali Ge, Yong Li, Liang Zong, Ju Gao
ABSTRACT
Progressive lung injury and pulmonary inflammation can be induced by an intraperitoneal injection of lipopolysaccharide (LPS). Interleukin-1β (IL-1β) is a key pro-inflammatory cytokine that can further exaggerate inflammation, which is cleaved and activated by the NALP3 inflammasome. Although the nuclear receptor Rev-erbα attenuates the level of LPS-induced pulmonary inflammation, the mechanism remains unclear. In this study, we investigated the influence of LPS-induced production of IL-1β and Rev-erbα on the development of lung inflammation. Herein, we demonstrate that Rev-erbα reduces IL-1β production and lung injury following an intraperitoneal injection of LPS, which is dependent on the NF-κB/NALP3 pathway. Thus, Rev-erbα is able to decrease the extent of acute lung injury by regulating IL-1β production. This mechanism may represent a potential novel therapeutic approach for lung injury.