MilliporeSigma
  • Assessment of mycotoxin exposure and risk characterization using occurrence data in foods and urinary biomarkers in Brazil.
Assessment of mycotoxin exposure and risk characterization using occurrence data in foods and urinary biomarkers in Brazil.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2019-03-31)
Larissa T Franco, Tânia Petta, George E Rottinghaus, Keliani Bordin, Gilmar A Gomes, Paula Alvito, Ricardo Assunção, Carlos A F Oliveira
ABSTRACT
This study aimed to assess the exposure of Brazilian residents (N = 86) from rural areas to multiple mycotoxins and characterize the associated risk in two sampling periods (SP) (April-May and December/2016). Mycotoxins in food and urine samples were determined by liquid chromatography coupled to tandem mass spectrometry. Mean probable daily intake (PDI) values based on occurrence data in foods in both SP varied from 0.007 to 0.013, 0.069 to 1.002, 0.119 to 0.321 and 0.013-0.156 μg kg-1 body weight (bw) day-1 for aflatoxins (AFs), deoxynivalenol (DON), fumonisins (FBs) and zearalenone (ZEN), respectively. Mean PDI values based on urinary biomarkers were 0.001, 84.914, 0.031, 0.377 and 0.002 μg kg-1 bw day-1 for AFB1, DON, ochratoxin A (OTA), FB1 and ZEN, respectively. Hazard quotient (HQ) calculated using food data revealed a potential health concern for ZEN in 2nd SP. HQ > 1 based on urinary biomarkers were observed for DON in the two SP. Although OTA was not detected in any food sample, the HQ based on urinary OTA levels was >1 in the 1st SP. Margin of exposure values for AF from food and urine data in the 1st SP were below 10,000, indicating potential health risks.