MilliporeSigma
  • DNA Methylation Inhibitor 5-Aza-2'-Deoxycytidine Modulates Endometrial Receptivity Through Upregulating HOXA10 Expression.
DNA Methylation Inhibitor 5-Aza-2'-Deoxycytidine Modulates Endometrial Receptivity Through Upregulating HOXA10 Expression.
Reproductive sciences (Thousand Oaks, Calif.) (2018-12-14)
Li Wang, Ya Jing Tan, Min Wang, Yi Fei Chen, Xin Yan Li
ABSTRACT
Endometrial receptivity is a critical factor for embryo implantation. A decrease in endometrial homeobox A10 (HOXA10) expression is associated with hypermethylation of its promoter and lower endometrial receptivity in animals and humans. 5-Aza-2'-deoxycytidine (AZA) is a DNA methyltransferase inhibitor. However, whether demethylation of the HOXA10 gene could increase the receptivity of the human endometrium remains unknown. Homeobox A10 promoter methylation was analyzed using bisulfite genomic sequencing polymerase chain reaction. Quantitative real time polymerase chain reaction and Western blotting were used to analyze the expression of HOXA10 and its downstream target genes (integrin subunit β 3 [ITGB3] and insulin growth factor binding protein 1 [IGFBP1]) in Ishikawa cells treated with or without AZA for 24 hours. Their protein expression was analyzed with or without HOXA10 siRNA treatment. The effect of AZA on embryo implantation was examined using a Jeg-3 spheroid-endometrial cell attachment assay. The percentage of methylated CpG islands in the HOXA10 promoter was 72.0% without AZA treatment. However, it was 38% and 35% in the 1 and 10 μM AZA treatment groups, respectively. 5-Aza-2'-deoxycytidine strongly induced the expression of HOXA10, ITGB3, and IGFBP1 messenger RNA and their protein expression. Homeobox A10 knockdown led to decreased expression of HOXA10, ITGB3, and IGFBP1, with or without AZA treatment. The attachment rate of Jeg-3 spheroids increased significantly from 82% (control) to 95% (AZA 1 μM) and 96% (AZA 10 μM) after AZA treatment. 5-Aza-2'-deoxycytidine could upregulate the expression of ITGB3 and IGFBP1 via HOXA10 upregulation, and upregulation of ITGB3 and IGFBP1 plays an important role in endometrial receptivity during implantation. 5-Aza-2'-deoxycytidine may improve endometrial receptivity by upregulating the expression of HOXA10.