MilliporeSigma
  • U-turn trajectories of magnetotactic cocci allow the study of the correlation between their magnetic moment, volume and velocity.
U-turn trajectories of magnetotactic cocci allow the study of the correlation between their magnetic moment, volume and velocity.
European biophysics journal : EBJ (2019-06-17)
Daniel Acosta-Avalos, Agnes Chacor de Figueiredo, Cassia Picanço Conceição, Jayane Julia Pereira da Silva, Kaio José Monteiro São Paulo Aguiar, Marciano de Lima Medeiros, Moacyr do Nascimento, Roger Duarte de Melo, Saulo Machado Moreira Sousa, Henrique Lins de Barros, Odivaldo Cambraia Alves, Fernanda Abreu
ABSTRACT
Magnetotactic bacteria are microorganisms that present intracellular chains of magnetic nanoparticles, the magnetosome chain. A challenge in the study of magnetotactic bacteria is the measurement of the magnetic moment associated with the magnetosome chain. Several techniques have been used to estimate the average magnetic moment of a population of magnetotactic bacteria, and others permit the measurement of the magnetic moment of individual bacteria. The U-turn technique allows the measurement of the individual magnetic moment and other parameters associated with the movement and magnetotaxis, such as the velocity and the orientation angle of the trajectory relative to the applied magnetic field. The aim of the present paper is to use the U-turn technique in a population of uncultured magnetotactic cocci to measure the magnetic moment, the volume, orientation angle and velocity for the same individuals. Our results showed that the magnetic moment is distributed in a log-normal distribution, with a mean value of 8.2 × 10-15 Am2 and median of 5.4 × 10-15 Am2. An estimate of the average magnetic moment using the average value of the orientation cosine produces a value similar to the median of the distribution and to the average magnetic moment obtained using transmission electron microscopy. A strong positive correlation is observed between the magnetic moment and the volume. There is no correlation between the magnetic moment and the orientation cosine and between the magnetic moment and the velocity. Those null correlations can be explained by our current understanding of magnetotaxis.