MilliporeSigma
DEL-1 promotes macrophage efferocytosis and clearance of inflammation.
Nature immunology (2018-11-21)
Ioannis Kourtzelis, Xiaofei Li, Ioannis Mitroulis, Daniel Grosser, Tetsuhiro Kajikawa, Baomei Wang, Michal Grzybek, Janusz von Renesse, Aleksander Czogalla, Maria Troullinaki, Anaisa Ferreira, Christian Doreth, Klara Ruppova, Lan-Sun Chen, Kavita Hosur, Jong-Hyung Lim, Kyoung-Jin Chung, Sylvia Grossklaus, Anne Kathrin Tausche, Leo A B Joosten, Niki M Moutsopoulos, Ben Wielockx, Antonio Castrillo, Jonathan M Korostoff, Ünal Coskun, George Hajishengallis, Triantafyllos Chavakis
ABSTRACT
Resolution of inflammation is essential for tissue homeostasis and represents a promising approach to inflammatory disorders. Here we found that developmental endothelial locus-1 (DEL-1), a secreted protein that inhibits leukocyte-endothelial adhesion and inflammation initiation, also functions as a non-redundant downstream effector in inflammation clearance. In human and mouse periodontitis, waning of inflammation was correlated with DEL-1 upregulation, whereas resolution of experimental periodontitis failed in DEL-1 deficiency. This concept was mechanistically substantiated in acute monosodium-urate-crystal-induced inflammation, where the pro-resolution function of DEL-1 was attributed to effective apoptotic neutrophil clearance (efferocytosis). DEL-1-mediated efferocytosis induced liver X receptor-dependent macrophage reprogramming to a pro-resolving phenotype and was required for optimal production of at least certain specific pro-resolving mediators. Experiments in transgenic mice with cell-specific overexpression of DEL-1 linked its anti-leukocyte-recruitment action to endothelial cell-derived DEL-1 and its efferocytic/pro-resolving action to macrophage-derived DEL-1. Thus, the compartmentalized expression of DEL-1 facilitates distinct homeostatic functions in an appropriate context that can be harnessed therapeutically.