MilliporeSigma
  • Effects of exercise and dietary protein sources on adiposity and insulin sensitivity in obese mice.
Effects of exercise and dietary protein sources on adiposity and insulin sensitivity in obese mice.
The Journal of nutritional biochemistry (2019-02-19)
Even Fjære, Lene Secher Myrmel, Ditte Olsen Lützhøft, Hanne Andersen, Jacob Bak Holm, Pia Kiilerich, Rita Hannisdal, Bjørn Liaset, Karsten Kristiansen, Lise Madsen
ABSTRACT
Low-fat diets and exercise are generally assumed to ameliorate obesity-related metabolic dysfunctions, but the importance of exercise vs. dietary changes is debated. Male C57BL/6J mice were fed a high-fat/high-sucrose (HF/HS) diet to induce obesity and then either maintained on the HF/HS or shifted to low-fat (LF) diets containing either salmon or entrecote. For each diet, half of the animals exercised voluntarily for 8 weeks. We determined body composition, glucose tolerance, insulin sensitivity and hepatic triacylglycerol levels. The microbiota composition in cecal and fecal samples was analyzed using 16S ribosomal RNA gene amplicon sequencing. Voluntary exercise improved insulin sensitivity but did not improve glucose tolerance. Voluntary exercise did not reduce adiposity in mice maintained on an HF/HS diet but enhanced LF-induced reduction in adiposity. Hepatic triacylglycerol levels were reduced by voluntary exercise in LF- but not HF/HS-fed mice. Voluntary exercise induced shifts in the cecal and fecal microbiota composition and functional potential in mice fed LF or HF/HS diets. Whereas voluntary exercise improved insulin sensitivity, a switch to an LF diet was the most important factor related to body weight and fat mass reduction.