MilliporeSigma
  • Induced Pluripotent Stem Cells and Their Use in Human Models of Disease and Development.
Induced Pluripotent Stem Cells and Their Use in Human Models of Disease and Development.
Physiological reviews (2018-10-18)
Peter Karagiannis, Kazutoshi Takahashi, Megumu Saito, Yoshinori Yoshida, Keisuke Okita, Akira Watanabe, Haruhisa Inoue, Jun K Yamashita, Masaya Todani, Masato Nakagawa, Mitsujiro Osawa, Yoshimi Yashiro, Shinya Yamanaka, Kenji Osafune
ABSTRACT
The discovery of somatic cell nuclear transfer proved that somatic cells can carry the same genetic code as the zygote, and that activating parts of this code are sufficient to reprogram the cell to an early developmental state. The discovery of induced pluripotent stem cells (iPSCs) nearly half a century later provided a molecular mechanism for the reprogramming. The initial creation of iPSCs was accomplished by the ectopic expression of four specific genes (OCT4, KLF4, SOX2, and c-Myc; OSKM). iPSCs have since been acquired from a wide range of cell types and a wide range of species, suggesting a universal molecular mechanism. Furthermore, cells have been reprogrammed to iPSCs using a myriad of methods, although OSKM remains the gold standard. The sources for iPSCs are abundant compared with those for other pluripotent stem cells; thus the use of iPSCs to model the development of tissues, organs, and other systems of the body is increasing. iPSCs also, through the reprogramming of patient samples, are being used to model diseases. Moreover, in the 10 years since the first report, human iPSCs are already the basis for new cell therapies and drug discovery that have reached clinical application. In this review, we examine the generation of iPSCs and their application to disease and development.