MilliporeSigma
  • PHF7 is a novel histone H2A E3 ligase prior to histone-to-protamine exchange during spermiogenesis.
PHF7 is a novel histone H2A E3 ligase prior to histone-to-protamine exchange during spermiogenesis.
Development (Cambridge, England) (2019-06-14)
Xiukun Wang, Jun-Yan Kang, Leixin Wei, Xiaogan Yang, Hongduo Sun, Suming Yang, Lei Lu, Meng Yan, Meizhu Bai, Yanyan Chen, Juanjuan Long, Na Li, Dangsheng Li, Jing Huang, Ming Lei, Zhen Shao, Wen Yuan, Erwei Zuo, Kehuan Lu, Mo-Fang Liu, Jinsong Li
ABSTRACT
Epigenetic regulation, including histone-to-protamine exchanges, controls spermiogenesis. However, the underlying mechanisms of this regulation are largely unknown. Here, we report that PHF7, a testis-specific PHD and RING finger domain-containing protein, is essential for histone-to-protamine exchange in mice. PHF7 is specifically expressed during spermiogenesis. PHF7 deletion results in male infertility due to aberrant histone retention and impaired protamine replacement in elongated spermatids. Mechanistically, PHF7 can simultaneously bind histone H2A and H3; its PHD domain, a histone code reader, can specifically bind H3K4me3/me2, and its RING domain, a histone writer, can ubiquitylate H2A. Thus, our study reveals that PHF7 is a novel E3 ligase that can specifically ubiquitylate H2A through binding H3K4me3/me2 prior to histone-to-protamine exchange.