MilliporeSigma
  • Association Study of ARMC9 Gene Variants with Vogt-Koyanagi-Harada Disease in Japanese Patients.
Association Study of ARMC9 Gene Variants with Vogt-Koyanagi-Harada Disease in Japanese Patients.
Ocular immunology and inflammation (2018-11-06)
Tomoko Ohno, Akira Meguro, Masaki Takeuchi, Takahiro Yamane, Takeshi Teshigawara, Nobuyoshi Kitaichi, Yukihiro Horie, Kenichi Namba, Shigeaki Ohno, Kumiko Nakao, Taiji Sakamoto, Tsutomu Sakai, Tadashi Nakano, Hiroshi Keino, Annabelle A Okada, Atsunobu Takeda, Takako Fukuhara, Hisashi Mashimo, Nobuyuki Ohguro, Shinichirou Oono, Hiroshi Enaida, Satoshi Okinami, Nobuhisa Mizuki
ABSTRACT
Purpose: To investigate whether variants in the ARMC9 gene encoding KU-MEL-1 are associated with Vogt-Koyanagi-Harada (VKH) disease in a Japanese population. Methods: We recruited 380 Japanese patients with VKH disease and 744 Japanese healthy controls to genotype seven single-nucleotide polymorphisms (SNPs) in ARMC9. We also performed imputation analysis of the ARMC9 region and 195 imputed SNPs were included in the statistical analysis. Results: We observed an increased frequency of the A allele of rs28690417 in patients compared with controls (P = 0.0097, odds ratio (OR) = 1.46). The A allele had a dominant effect on VKH disease risk (P = 0.011, OR = 1.51). However, these significant differences disappeared after Bonferroni correction (corrected P > 0.05). The remaining 201 SNPs did not show any significant association with disease risk. Conclusions: Our study suggests that ARMC9 variants do not play a critical role in the development of VKH disease.