MilliporeSigma
  • Sirt1-hypoxia-inducible factor-1α interaction is a key mediator of tubulointerstitial damage in the aged kidney.
Sirt1-hypoxia-inducible factor-1α interaction is a key mediator of tubulointerstitial damage in the aged kidney.
Aging cell (2019-01-08)
Dong Ryeol Ryu, Mi Ra Yu, Kyoung Hye Kong, Hyoungnae Kim, Soon Hyo Kwon, Jin Seok Jeon, Dong Cheol Han, Hyunjin Noh
ABSTRACT
Although it is known that the expression and activity of sirtuin 1 (Sirt1) decrease in the aged kidney, the role of interaction between Sirt1 and hypoxia-inducible factor (HIF)-1α is largely unknown. In this study, we investigated whether HIF-1α could be a deacetylation target of Sirt1 and the effect of their interaction on age-associated renal injury. Five-week-old (young) and 24-month-old (old) C57Bl/6J mice were assessed for their age-associated changes. Kidneys from aged mice showed increased infiltration of CD68-positive macrophages, higher expression of extracellular matrix (ECM) proteins, and more apoptosis than young controls. They also showed decreased Sirt1 expression along with increased acetylated HIF-1α. The level of Bcl-2/adenovirus E1B-interacting protein 3, carbonic anhydrase 9, Snail, and transforming growth factor-β1, which are regulated by HIF-1α, was significantly higher in aged mice suggesting that HIF-1α activity was increased. In HK-2 cells, Sirt1 inhibitor sirtinol and siRNA-mediated knockdown of Sirt1 enhanced apoptosis and ECM accumulation. During hypoxia, Sirt1 was down-regulated, which allowed the acetylation and activation of HIF-1α. Resveratrol, a Sirt1 activator, effectively prevented hypoxia-induced production of ECM proteins, mitochondrial damage, reactive oxygen species generation, and apoptosis. The inhibition of HIF-1α activity by Sirt1-induced deacetylation of HIF-1α was confirmed by Sirt1 overexpression under hypoxic conditions and by resveratrol treatment or Sirt1 overexpression in HIF-1α-transfected HK-2 cells. Finally, we confirmed that chronic activation of HIF-1α promoted apoptosis and fibrosis, using tubular cell-specific HIF-1α transgenic mice. Taken together, our data suggest that Sirt1-induced deacetylation of HIF-1α may have protective effects against tubulointerstitial damage in aged kidney.