MilliporeSigma
  • Effect of Thickness of Ceramic Veneers on Color Stability and Bond Strength of Resin Luting Cements Containing Alternative Photoinitiators.
Effect of Thickness of Ceramic Veneers on Color Stability and Bond Strength of Resin Luting Cements Containing Alternative Photoinitiators.
The journal of adhesive dentistry (2019-02-26)
Mauricio Castellanos, Alex Jose Delgado, Mario Alexandre Coelho Sinhoreti, Dayane Carvalho Ramos Salles de Oliveira, Nader Abdulhameed, Saulo Geraldeli, Jean-François Roulet
ABSTRACT
To evaluate the light transmittance of ceramic veneers of different thicknesses and verify their influence on the degree of conversion, color stability, and dentin bond strength of light-curing resin cements containing different photoinitiator systems. Experimental resin cements were fabricated containing camphorquinone and amine (CQ-amine), TPO, Ivocerin (IVO), or TPO and Ivocerin (TPO-IVO). All photoinitiators were characterized by UV-Vis spectrophotometry. Disk-shaped lithium disilicate ceramic specimens that were 0.4, 0.7, and 1.5 mm in thickness were prepared using IPS e.max Press (Ivoclar Vivadent, shade LT/A2). Light transmittance through each specimen was measured using spectrophotometry. Specimens of each cement (n = 10) were made in a custom-designed mold and were light cured through each glass-ceramic disk using a multiwave LED (Bluephase G2, Ivoclar Vivadent). CS was evaluated using spectrophotometry before and after artificial aging with UV light. DC was evaluated using FTIR-spectroscopy. Dentin µSBS was evaluated using 0.75-mm-thick specimens that were light cured under the same protocol (n = 10). All data were submitted to two-way ANOVA and Tukey's test (α = 0.05; β = 0.2). CQ-amine cements showed the highest color changes (p < 0.05) due to increased yellowing when compared to the amine-free cements (p < 0.05). However, all cements showed a significant color change after aging when cured through ceramics up to 1.5 mm thick (p < 0.05). The TPO-IVO cement showed the highest DC and the IVO cement showed a similar DC when compared to the CQ-amine cement. The TPO cement presented the lowest DC (p = 0.0377). No differences in mean dentin µSBS were found among the cements, except for the TPO cement, which presented a lower mean dentin µSBS (p = 0.0277). Amine-free cements containing Ivocerin and TPO seem to be a better alternative to CQ-amine cements, while not reducing either DC or dentin µSBS of amine-free cements. However, CQ-amine and amine-free cements still seem to change color over time.