MilliporeSigma
  • Preparation and Thermal Evaluation of Novel Polyimide Protective Coatings for Quartz Capillary Chromatographic Columns Operated over 320 °C for High-Temperature Gas Chromatography Analysis.
Preparation and Thermal Evaluation of Novel Polyimide Protective Coatings for Quartz Capillary Chromatographic Columns Operated over 320 °C for High-Temperature Gas Chromatography Analysis.
Polymers (2019-06-05)
Meng-Ge Huangfu, Yan Zhang, Xin-Ling Zhang, Jin-Gang Liu, Ying-Cong Liu, Yi-Dan Guo, Qing-Yuan Huang, Xiu-Min Zhang
ABSTRACT
Protection of intrinsically brittle quartz chromatographic columns (CCs) from breakage or property deterioration in gas chromatography (GC) analysis has become an important research topic regarding high-temperature GC techniques. Polyimide (PI) has proved to be the most suitable protective coating for quartz CCs. In the current research, a series of novel high-temperature-resistant PI coatings for quartz CCs operated over 320 °C have been successfully prepared. For this purpose, the aromatic diamine with a rigid skeleton structure 2-(4-aminophenyl)-5-aminobenzimidazole (APBI) was copolymerized with two aromatic dianhydrides-3,3',4,4'-benzophenotetracarboxylic acid dianhydride (BTDA) and 4,4'-oxydiphthalic anhydride (ODPA)-and an aromatic diamine with flexible ether linkages-4,4'-oxydianiline (ODA)-by a two-step polymerization procedure via soluble poly(amic acid) (PAA) precursors, followed by thermal imidization at elevated temperatures. The developed PI coatings exhibited good comprehensive properties, including glass transition temperatures (Tg) as high as 346.9 °C, measured by dynamic mechanical analysis (DMA), and coefficients of linear thermal expansion (CTEs) as low as 24.6 × 10-6/K in the range of 50-300 °C. In addition, the PI coatings exhibited good adhesion to the fused quartz capillary columns. No cracking, delamination, warpage, or other failures occurred during the 100-cycle thermal shock test in the range of 25-320 °C.