MilliporeSigma
  • Differential expression and signaling of the human histamine H3 receptor isoforms of 445 and 365 amino acids expressed in human neuroblastoma SH-SY5Y cells.
Differential expression and signaling of the human histamine H3 receptor isoforms of 445 and 365 amino acids expressed in human neuroblastoma SH-SY5Y cells.
Journal of receptor and signal transduction research (2018-03-21)
Gustavo Nieto-Alamilla, Juan Escamilla-Sánchez, María-Cristina López-Méndez, Anayansi Molina-Hernández, Agustín Guerrero-Hernández, José-Antonio Arias-Montaño
ABSTRACT
In stably-transfected human neuroblastoma SH-SY5Y cells, we have compared the effect of activating two isoforms of 445 and 365 amino acids of the human histamine H3 receptor (hH3R445 and hH3R365) on [35S]-GTPγS binding, forskolin-induced cAMP formation, depolarization-induced increase in the intracellular concentration of Ca2+ ions ([Ca2+]i) and depolarization-evoked [3 H]-dopamine release. Maximal specific binding (Bmax) of [3 H]-N-methyl-histamine to cell membranes was 953 ± 204 and 555 ± 140 fmol/mg protein for SH-SY5Y-hH3R445 and SH-SY5Y-hH3R365 cells, respectively, with similar dissociation constants (Kd, 0.86 nM and 0.81 nM). The mRNA of the hH3R365 isoform was 40.9 ± 7.9% of the hH3R445 isoform. No differences in receptor affinity were found for the H3R ligands histamine, immepip, (R)(-)-α-methylhistamine (RAMH), A-331440, clobenpropit and ciproxifan. Both the stimulation of [35S]-GTPγS binding and the inhibition of forskolin-stimulated cAMP accumulation by the agonist RAMH were significantly larger in SH-SY5Y-hH3R445 cells ([35S]-GTPγS binding, 158.1 ± 7.5% versus 136.5 ± 3.6% for SH-SY5Y-hH3R365 cells; cAMP accumulation, -74.0 ± 4.9% versus -43.5 ± 5.3%), with no significant effect on agonist potency. In contrast, there were no differences in the efficacy and potency of RAMH to inhibit [3 H]-dopamine release evoked by 100 mM K+ (-18.9 ± 3.0% and -20.5 ± 3.3%, for SH-SY5Y-hH3R445 and SH-SY5Y-hH3R365 cells), or the inhibition of depolarization-induced increase in [Ca2+]i (S2/S1 ratios: parental cells 0.967 ± 0.069, SH-SY5Y-hH3R445 cells 0.639 ± 0.049, SH-SY5Y-hH3R365 cells 0.737 ± 0.045). These results indicate that in SH-SY5Y cells, hH3R445 and hH3R365 isoforms regulate in a differential manner the signaling pathways triggered by receptor activation.