MilliporeSigma
  • Label-free quantitation of glycated hemoglobin in single red blood cells by transient absorption microscopy and phasor analysis.
Label-free quantitation of glycated hemoglobin in single red blood cells by transient absorption microscopy and phasor analysis.
Science advances (2019-05-17)
Pu-Ting Dong, Haonan Lin, Kai-Chih Huang, Ji-Xin Cheng
ABSTRACT
As a stable and accurate biomarker, glycated hemoglobin (HbA1c) is clinically used to diagnose diabetes with a threshold of 6.5% among total hemoglobin (Hb). Current methods such as boronate affinity chromatography involve complex processing of large-volume blood samples. Moreover, these methods cannot measure HbA1c fraction at single-red blood cell (RBC) level, thus unable to separate the contribution from other factors such as RBC lifetime. Here, we demonstrate a spectroscopic transient absorption imaging approach that is able to differentiate HbA1c from Hb on the basis of their distinct excited-state dynamics. HbA1c fraction inside a single RBC is derived quantitatively through phasor analysis. HbA1c fraction distribution of diabetic blood is apparently different from that of healthy blood. A mathematical model is developed to derive the long-term blood glucose concentration. Our technology provides a unique way to study heme modification and to derive clinically important information void of bloodstream glucose fluctuation.