MilliporeSigma
  • MTHFD1 interaction with BRD4 links folate metabolism to transcriptional regulation.
MTHFD1 interaction with BRD4 links folate metabolism to transcriptional regulation.
Nature genetics (2019-05-28)
Sara Sdelci, André F Rendeiro, Philipp Rathert, Wanhui You, Jung-Ming G Lin, Anna Ringler, Gerald Hofstätter, Herwig P Moll, Bettina Gürtl, Matthias Farlik, Sandra Schick, Freya Klepsch, Matthew Oldach, Pisanu Buphamalai, Fiorella Schischlik, Peter Májek, Katja Parapatics, Christian Schmidl, Michael Schuster, Thomas Penz, Dennis L Buckley, Otto Hudecz, Richard Imre, Shuang-Yan Wang, Hans Michael Maric, Robert Kralovics, Keiryn L Bennett, Andre C Müller, Karl Mechtler, Jörg Menche, James E Bradner, Georg E Winter, Kristaps Klavins, Emilio Casanova, Christoph Bock, Johannes Zuber, Stefan Kubicek
ABSTRACT
The histone acetyl reader bromodomain-containing protein 4 (BRD4) is an important regulator of chromatin structure and transcription, yet factors modulating its activity have remained elusive. Here we describe two complementary screens for genetic and physical interactors of BRD4, which converge on the folate pathway enzyme MTHFD1 (methylenetetrahydrofolate dehydrogenase, cyclohydrolase and formyltetrahydrofolate synthetase 1). We show that a fraction of MTHFD1 resides in the nucleus, where it is recruited to distinct genomic loci by direct interaction with BRD4. Inhibition of either BRD4 or MTHFD1 results in similar changes in nuclear metabolite composition and gene expression; pharmacological inhibitors of the two pathways synergize to impair cancer cell viability in vitro and in vivo. Our finding that MTHFD1 and other metabolic enzymes are chromatin associated suggests a direct role for nuclear metabolism in the control of gene expression.