MilliporeSigma
  • Piperine, a component of black pepper, inhibits adipogenesis by antagonizing PPARγ activity in 3T3-L1 cells.
Piperine, a component of black pepper, inhibits adipogenesis by antagonizing PPARγ activity in 3T3-L1 cells.
Journal of agricultural and food chemistry (2012-04-03)
Ui-Hyun Park, Hong-Suk Jeong, Eun-Young Jo, Taesun Park, Seung Kew Yoon, Eun-Joo Kim, Ji-Cheon Jeong, Soo-Jong Um
ABSTRACT
This study investigated the antiadipogenic activity of black pepper extract and its constituent piperine in 3T3-L1 preadipocytes as well as the underlying molecular mechanisms. Both black pepper extract and piperine, without affecting cytotoxicity, strongly inhibited the adipocyte differentiation of 3T3-L1 cells. The mRNA expression of the master adipogenic transcription factors, PPARγ, SREBP-1c, and C/EBPβ, was markedly decreased. Intriguingly, mRNA levels of PPARγ target genes were also down-regulated. Moreover, a luciferase reporter assay indicated that pipierine significantly represses the rosiglitazone-induced PPARγ transcriptional activity. Finally, GST-pull down assays demonstrated that piperine disrupts the rosiglitazone-dependent interaction between PPARγ and coactivator CBP. Genome-wide analysis using microarray further supports the role of piperine in regulating genes associated with lipid metabolism. Overall, these results suggest that piperine, a major component of black pepper, attenuates fat cell differentiation by down-regulating PPARγ activity as well as suppressing PPARγ expression, thus leading to potential treatment for obesity-related diseases.
Sigma-Aldrich
Piperine, ≥95%, FG