MilliporeSigma
  • Using supramolecular hydrogels to discover the interactions between proteins and molecular nanofibers of small molecules.
Using supramolecular hydrogels to discover the interactions between proteins and molecular nanofibers of small molecules.
Chemical communications (Cambridge, England) (2012-07-18)
Yuan Gao, Marcus J C Long, Junfeng Shi, Lizbeth Hedstrom, Bing Xu
ABSTRACT
Here we report the first example of the use of supramolecular hydrogels to discover the protein targets of aggregates of small molecules.