MilliporeSigma
  • The medicinal plant goldenseal is a natural LDL-lowering agent with multiple bioactive components and new action mechanisms.
The medicinal plant goldenseal is a natural LDL-lowering agent with multiple bioactive components and new action mechanisms.
Journal of lipid research (2006-08-04)
Parveen Abidi, Wei Chen, Fredric B Kraemer, Hai Li, Jingwen Liu
ABSTRACT
Our previous studies have identified berberine (BBR), an alkaloid isolated from the Chinese herb huanglian, as a unique cholesterol-lowering drug that upregulates hepatic low density lipoprotein receptor (LDLR) expression through a mechanism of mRNA stabilization. Here, we demonstrate that the root extract of goldenseal, a BBR-containing medicinal plant, is highly effective in upregulation of liver LDLR expression in HepG2 cells and in reducing plasma cholesterol and low density lipoprotein cholesterol (LDL-c) in hyperlipidemic hamsters, with greater activities than the pure compound BBR. By conducting bioassay-driven semipurifications, we demonstrate that the higher potency of goldenseal is achieved through concerted actions of multiple bioactive compounds in addition to BBR. We identify canadine (CND) and two other constituents of goldenseal as new upregulators of LDLR expression. We further show that the activity of BBR on LDLR expression is attenuated by multiple drug resistance-1 (MDR1)-mediated efflux from liver cells, whereas CND is resistant to MDR1. This finding defines a molecular mechanism for the higher activity of CND than BBR. We also provide substantial evidence to show that goldenseal contains natural MDR1 antagonist(s) that accentuate the upregulatory effect of BBR on LDLR mRNA expression. These new findings identify goldenseal as a natural LDL-c-lowering agent, and our studies provide a molecular basis for the mechanisms of action.
Sigma-Aldrich
MDR1 Efflux Assay, The Multidrug Resistance Direct Dye Efflux Assay Kit includes two of the best characterized & most commonly used multidrug resistance ABC transporter substrates, DiOC2(3) & rhodamine 123.
Pricing and availability is not currently available.