MilliporeSigma
  • Direct effects of polymyxin B on human dendritic cells maturation. The role of IkappaB-alpha/NF-kappaB and ERK1/2 pathways and adhesion.
Direct effects of polymyxin B on human dendritic cells maturation. The role of IkappaB-alpha/NF-kappaB and ERK1/2 pathways and adhesion.
The Journal of biological chemistry (2005-01-27)
Barbara Valentinis, Alessandro Bianchi, Dan Zhou, Arcadi Cipponi, Federica Catalanotti, Vincenzo Russo, Catia Traversari
ABSTRACT
Polymyxin B is a lipopolysaccharide binding antibiotic used to inactivate potential lipopolysaccharide contaminations when evaluating the activity of different agents on innate immune cells. We report that polymyxin B is able to induce directly in monocyte-derived human dendritic cells (DCs) several functional and molecular modifications characteristic of DCs undergoing a maturation process. DCs incubated with polymyxin B up-regulate the expression of HLA class I and II, the co-stimulatory CD86 molecule, and show an increase in the fraction of adherent cells at short time, which persist at 48 h of incubation. Adhesion to the plate was required for the polymyxin B-induced DCs maturation. A transient activation of IkappaB-alpha/NF-kappaB and ERK1/2 pathways at short time and a further ERK1/2 activation at long term were also detected. Neither up-regulation of the maturation marker CD83 nor activation of p38 nor induction of cytokines secretion was observed in DCs treated with polymyxin B. We demonstrated that inhibition of IkappaB-alpha/NF-kappaB pathway abolishes polymyxin B effects. ERK1/2 inhibition instead allowed DCs treated with polymyxin B to progress in their maturation process as revealed by the increased up-regulation of the CD83 co-stimulatory molecules, the activation of p38, and the reduced adhesion to culture plates at 48 h of incubation. Our results indicate that polymyxin B induces a partial maturation of human DCs through increased adhesion to a substrate and activation of the IkappaB-alpha/NF-kappaB pathway. The increased ERK1/2 activation observed, even though correlating with the initial phases of the maturation process, actually inhibits the occurrence of full maturation.
Sigma-Aldrich
Polymyxin B solution, 20 mg/mL in H2O