MilliporeSigma
  • Steady-state plasma and intrapulmonary concentrations of levofloxacin and ciprofloxacin in healthy adult subjects.
Steady-state plasma and intrapulmonary concentrations of levofloxacin and ciprofloxacin in healthy adult subjects.
Chest (2001-04-11)
M H Gotfried, L H Danziger, K A Rodvold
ABSTRACT
To determine the steady-state plasma, epithelial lining fluid (ELF), and alveolar macrophage (AM) concentrations of levofloxacin and ciprofloxacin. Multiple-dose, open-label, randomized pharmacokinetic study. Thirty-six healthy, nonsmoking adult subjects were randomized either to oral levofloxacin, 500 or 750 mg once daily for five doses, or ciprofloxacin, 500 mg q12h for nine doses. Venipuncture, bronchoscopy, and BAL were performed in each subject at 4 h, 12 h, or 24 h after the last administered dose of antibiotic. Mean plasma concentrations of levofloxacin and ciprofloxacin were similar to those previously reported. For once-daily dosing of levofloxacin, 500 mg, the mean (+/- SD) steady-state concentrations at 4 h, 12 h, and 24 h in ELF were 9.9 +/- 2.7 microg/mL, 6.5 +/- 2.5 microg/mL, and 0.7 +/- 0.4 microg/mL, respectively; AM concentrations were 97.9 +/- 80.0 microg/mL, 36.7 +/- 23.4 microg/mL, and 13.8 +/- 16.0 microg/mL, respectively. For levofloxacin, 750 mg, the mean steady-state concentrations in ELF were 22.1 +/- 14.9 microg/mL, 9.2 +/- 5.3 microg/mL, and 1.5 +/- 0.8 microg/mL, respectively; AM concentrations were 105.1 +/- 65.5 microg/mL, 36.2 +/- 26.1 microg/mL, and 15.1 +/- 2.0 microg/mL, respectively. The concentrations of ciprofloxacin at 4 h and 12 h in ELF were 1.9 +/- 0.9 microg/mL and 0.4 +/- 0.1 microg/mL, respectively; AM concentrations were 34.9 +/- 23.2 microg/mL and 6.8 +/- 5.9 microg/mL, respectively. The differences in the ELF concentrations of the two levofloxacin groups vs those of the ciprofloxacin group were significant (p < 0.05) at each sampling time. Levofloxacin was more extensively distributed into intrapulmonary compartments than ciprofloxacin and achieved significantly higher steady-state concentrations in plasma and ELF during the 24 h after drug administration.
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)