MilliporeSigma
  • Functionalized Phenylbenzamides Inhibit Aquaporin-4 Reducing Cerebral Edema and Improving Outcome in Two Models of CNS Injury.
Functionalized Phenylbenzamides Inhibit Aquaporin-4 Reducing Cerebral Edema and Improving Outcome in Two Models of CNS Injury.
Neuroscience (2019-02-10)
George W Farr, Christopher H Hall, Susan M Farr, Ramon Wade, Joshua M Detzel, Amielia G Adams, Jasen M Buch, Derek L Beahm, Christopher A Flask, Kui Xu, Joseph C LaManna, Paul R McGuirk, Walter F Boron, Marc F Pelletier
ABSTRACT
Cerebral edema in ischemic stroke can lead to increased intracranial pressure, reduced cerebral blood flow and neuronal death. Unfortunately, current therapies for cerebral edema are either ineffective or highly invasive. During the development of cytotoxic and subsequent ionic cerebral edema water enters the brain by moving across an intact blood brain barrier and through aquaporin-4 (AQP4) at astrocyte endfeet. Using AQP4-expressing cells, we screened small molecule libraries for inhibitors that reduce AQP4-mediated water permeability. Additional functional assays were used to validate AQP4 inhibition and identified a promising structural series for medicinal chemistry. These efforts improved potency and revealed a compound we designated AER-270, N-[3,5-bis (trifluoromethyl)phenyl]-5-chloro-2-hydroxybenzamide. AER-270 and a prodrug with enhanced solubility, AER-271 2-{[3,5-Bis(trifluoromethyl) phenyl]carbamoyl}-4-chlorophenyl dihydrogen phosphate, improved neurological outcome and reduced swelling in two models of CNS injury complicated by cerebral edema: water intoxication and ischemic stroke modeled by middle cerebral artery occlusion.
Sigma-Aldrich
AER-271, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
IMD-0354, ≥98% (HPLC), solid
Pricing and availability is not currently available.