MilliporeSigma
  • A Small Molecule Targeting the Transmembrane Domain of Death Receptor p75NTR Induces Melanoma Cell Death and Reduces Tumor Growth.
A Small Molecule Targeting the Transmembrane Domain of Death Receptor p75NTR Induces Melanoma Cell Death and Reduces Tumor Growth.
Cell chemical biology (2018-10-09)
Eddy T H Goh, Zhi Lin, Bo Young Ahn, Vanessa Lopes-Rodrigues, Ngoc Ha Dang, Shuhailah Salim, Bryan Berger, Brian Dymock, Donna L Senger, Carlos F Ibáñez
ABSTRACT
Small molecules offer powerful ways to alter protein function. However, most proteins in the human proteome lack small-molecule probes, including the large class of non-catalytic transmembrane receptors, such as death receptors. We hypothesized that small molecules targeting the interfaces between transmembrane domains (TMDs) in receptor complexes may induce conformational changes that alter receptor function. Applying this concept in a screening assay, we identified a compound targeting the TMD of death receptor p75NTR that induced profound conformational changes and receptor activity. The compound triggered apoptotic cell death dependent on p75NTR and JNK activity in neurons and melanoma cells, and inhibited tumor growth in a melanoma mouse model. Due to their small size and crucial role in receptor activation, TMDs represent attractive targets for small-molecule manipulation of receptor function.
Sigma-Aldrich
NSC49652, ≥98% (HPLC)