MilliporeSigma
  • The 8-amino-7-oxopelargonate synthase from Bacillus sphaericus. Purification and preliminary characterization of the cloned enzyme overproduced in Escherichia coli.
The 8-amino-7-oxopelargonate synthase from Bacillus sphaericus. Purification and preliminary characterization of the cloned enzyme overproduced in Escherichia coli.
The Biochemical journal (1992-04-15)
O Ploux, A Marquet
ABSTRACT
The 8-amino-7-oxopelargonate synthase [6-carboxyhexanoyl-CoA:L-alanine carboxyhexanoyltransferase (decarboxylating); EC 2.3.1.47] from Bacillus sphaericus involved in biotin biosynthesis was purified from an Escherichia coli overproducing strain. The purification afforded an electrophoretically homogeneous enzyme with a specific activity of 0.67 unit/mg. The purified enzyme is a monomer of 41 kDa. N-Terminal sequencing of the first 14 amino acid residues showed complete agreement with the predicted sequence from the bioF gene. The pure enzyme showed the characteristic absorption band (425 nm) of pyridoxal 5'-phosphate-dependent enzymes. Furthermore, the holoenzyme was resolved during an affinity step yielding the inactive apoenzyme, which recovered activity and the 425 nm-absorption band on dialysis against pyridoxal 5'-phosphate. Km values for L-alanine and pimeloyl-CoA were respectively 3 mM and 1 microM.
Sigma-Aldrich
Phenyl-Sepharose CL-4B, extent of labeling: ~40 μmol per mL