MilliporeSigma
  • A beta 3 integrin mutation abolishes ligand binding and alters divalent cation-dependent conformation.
A beta 3 integrin mutation abolishes ligand binding and alters divalent cation-dependent conformation.
Science (New York, N.Y.) (1990-08-24)
J C Loftus, T E O'Toole, E F Plow, A Glass, A L Frelinger, M H Ginsberg
ABSTRACT
The ligand-binding function of integrin adhesion receptors depends on divalent cations. A mutant alpha IIb beta 3 integrin (platelet gpIIb/IIIa) that lacks ligand recognition shows immunologic evidence of a perturbed interaction with divalent cations. This was found to be caused by a G----T mutation that resulted in an Asp119----Tyr119 substitution in the beta 3 subunit. This residue is proximal to bound ligand and is in a conserved region among integrins that are enriched in oxygenated residues. The spacing of these residues aligns with the calcium-binding residues in EF hand proteins, suggesting interaction with receptor-bound divalent cation as a mechanism of ligand binding common to all integrins.