MilliporeSigma
  • Effects of manidipine vs. amlodipine on intrarenal haemodynamics in patients with arterial hypertension.
Effects of manidipine vs. amlodipine on intrarenal haemodynamics in patients with arterial hypertension.
British journal of clinical pharmacology (2012-12-18)
Christian Ott, Markus P Schneider, Ulrike Raff, Martin Ritt, Kristina Striepe, Marco Alberici, Roland E Schmieder
ABSTRACT
Intraglomerular pressure is one of the main drivers of progression of renal failure. Experimental data suggest that there are important differences between calcium channel blockers (CCBs) in their renal haemodynamic effects: manidipine reduces, whereas amlodipine increases intraglomerular pressure. The aim of this study was to investigate the effects of manidipine and amlodipine treatment on intragomerular pressure (P(glom)) in patients with mild to moderate essential hypertension. In this randomized, double-blind, parallel group study, hypertensive patients were randomly assigned to receive manidipine 20 mg (n = 54) or amlodipine 10 mg (n = 50) for 4 weeks. Renal plasma flow (RPF) and glomerular filtration rate (GFR) were determined by constant-infusion input-clearance technique with p-aminohippurate (PAH) and inulin. P(glom) and resistances of the afferent (R(A)) and efferent (R(E)) arterioles were calculated according to the model established by Gomez. P(glom) did not change in the manidipine group (P = 0.951), whereas a significant increase occurred in the amlodipine group (P = 0.009). There was a significant difference in the change of P(glom) by 1.2 mmHg between the manidipine and amlodipine group (P = 0.042). In both treatment arms, R(A) was reduced (manidipine P = 0.018; amlodipine P < 0.001). The reduction of R(A) was significantly more pronounced with amlodipine compared with manidipine treatment (P < 0.001). R(E) increased in both treatment arms (manidipine P = 0.012; amlodipine P = 0.002), with no difference between the treatment arms. Both CCBs significantly reduced systolic and diastolic blood pressure (BP) (both P < 0.001). However, amlodipine treatment resulted in a significantly greater decrease of BP compared with manidipine (P < 0.001). In accordance with experimental data after antihypertensive treatment of 4 weeks, intraglomerular pressure was significantly lower with the CCB manidipine than with amlodipine, resulting and explaining their disparate effects on albuminuria.
Sigma-Aldrich
Manidipine hydrochloride, ≥98% (HPLC)