MilliporeSigma
  • A phase 1 study of romidepsin and pralatrexate reveals marked activity in relapsed and refractory T-cell lymphoma.
A phase 1 study of romidepsin and pralatrexate reveals marked activity in relapsed and refractory T-cell lymphoma.
Blood (2017-11-17)
Jennifer E Amengual, Renee Lichtenstein, Jennifer Lue, Ahmed Sawas, Changchun Deng, Emily Lichtenstein, Karen Khan, Laine Atkins, Aishling Rada, Hye A Kim, Codruta Chiuzan, Matko Kalac, Enrica Marchi, Lorenzo Falchi, Mark A Francescone, Lawrence Schwartz, Serge Cremers, Owen A O'Connor
ABSTRACT
Peripheral T-cell lymphomas (PTCL) are a group of rare malignancies characterized by chemotherapy resistance and poor prognosis. Romidepsin and pralatrexate were approved by the US Food and Drug Administration for patients with relapsed/refractory PTCL, exhibiting response rates of 25% and 29% respectively. Based on synergy in preclinical models of PTCL, we initiated a phase 1 study of pralatrexate plus romidepsin in patients with relapsed/refractory lymphoma. This was a single institution dose-escalation study of pralatrexate plus romidepsin designed to determine the dose-limiting toxicities (DLTs), maximum tolerated dose, pharmacokinetic profile, and response rates. Patients were treated with pralatrexate (10 to 25 mg/m2) and romidepsin (12 to 14 mg/m2) on 1 of 3 schedules: every week × 3 every 28 days, every week × 2 every 21 days, and every other week every 28 days. Treatment continued until progression, withdrawal of consent, or medical necessity. Twenty-nine patients were enrolled and evaluable for toxicity. Coadministration of pralatrexate and romidepsin was safe, well tolerated, with 3 DLTs across all schedules (grade 3 oral mucositis × 2; grade 4 sepsis × 1). The recommended phase 2 dose was defined as pralatrexate 25 mg/m2 and romidepsin 12 mg/m2 every other week. Twenty-three patients were evaluable for response. The overall response rate was 57% (13/23) across all patients and 71% (10/14) in PTCL. The phase 1 study of pralatrexate plus romidepsin resulted in a high response rate in patients with previously treated PTCL. A phase 2 study in PTCL will determine the efficacy of the combination. This trial was registered at www.clinicaltrials.gov as #NCT01947140.
Sigma-Aldrich
Pralatrexate, ≥98% (HPLC)