MilliporeSigma
  • Mitogen-activated protein kinase phosphorylates and regulates the HIV-1 Vif protein.
Mitogen-activated protein kinase phosphorylates and regulates the HIV-1 Vif protein.
The Journal of biological chemistry (1998-10-29)
X Yang, D Gabuzda
ABSTRACT
The human immunodeficiency virus type 1 (HIV-1) Vif protein plays a critical role in virus replication and infectivity. Here we show that Vif is phosphorylated and regulated by p44/42 mitogen-activated protein kinase (MAPK). Vif phosphorylation by MAPK was demonstrated in vitro as well as in vivo and was shown to occur on serine and threonine residues. Two-dimensional tryptic phosphopeptide mapping indicated that Vif is phosphorylated by MAPK on the same sites in vitro and in vivo. Radioactive peptide sequencing identified two phosphorylation sites, Thr96 and Ser165. These phosphorylation sites do not correspond to the known optimum consensus sequences for phosphorylation by MAPK (PX(S/T)P) nor to the minimum consensus sequence ((S/T)P), indicating that MAPK can phosphorylate proteins at sites other than those containing the PX(S/T)P or (S/T)P motifs. Synthetic Vif peptides corresponding to the local sequences of the phosphorylation sites were not phosphorylated by MAPK, suggesting that recognition of these sites by MAPK is likely to require structural determinants outside the phosphorylation site. Mutations of the Thr96 site, which is conserved among Vif sequences from HIV-1, HIV-2, and SIV, resulted in significant loss of Vif activity and inhibition of HIV-1 replication. These results suggest that MAPK plays a direct role in regulating HIV-1 replication and infectivity by phosphorylating Vif and identify a novel mechanism for activation of HIV-1 replication by mitogens and other extracellular stimuli.
Sigma-Aldrich
MAP Kinase 2/Erk2 Protein, active, mouse, 10 µg, Active, N-terminal GST-tagged, recombinant MAP Kinase 2, expressed in E. coli cells, activated using MEK1 (Catalog #14-429). For use in Kinase Assays.