MilliporeSigma
  • Protein from red cabbage (Brassica oleracea) seeds with antifungal, antibacterial, and anticancer activities.
Protein from red cabbage (Brassica oleracea) seeds with antifungal, antibacterial, and anticancer activities.
Journal of agricultural and food chemistry (2011-08-13)
Xiu-Juan Ye, Tzi-Bun Ng, Zu-Jian Wu, Lian-Hui Xie, Evandro-Fei Fang, Jack-Ho Wong, Wen-Liang Pan, Sze-Stephen-Cho Wing, Yan-Bo Zhang
ABSTRACT
A 30 kDa antifungal protein was purified from red cabbage ( Brassica oleracea ) seeds. It exhibited a molecular mass and N-terminal amino acid sequence disinct from those of previously isolated Brassica antifungal proteins. The protocol used entailed ion exchange chromatography on Q-Sepharose and SP-Sepharose followed by fast protein liquid chromatography on Mono S. The protein hindered mycelial growth in Mycosphaerella arachidicola (with an IC50=5 μM), Setospaeria turcica, and Bipolaris maydis. It also inhibited the yeast Candida albicans with an IC50=96 μM. It exerted its antifungal action by permeabilizing the fungal membrane as evidenced by staining with Sytox green. The antifungal activity was stable from pH 3 to 11 and from 0 to 65 °C. It manifested antibacterial activity against Pseudomonas aeruginosa (IC50=53 μM). Furthermore, after 48 h of culture, it suppressed proliferation of nasopharyngeal cancer and hepatoma cells with IC50=50 and 90 μM, respectively.